Stampa…
Anti-SEPN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornitore: United States Biological
USBI041554-FITCEA 852 EUR
USBI041554-FITC
Anti-SEPN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SEPN1 (selenoprotein N, 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human