Stampa…
Anti-SENP6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornitore: United States Biological
USBI041552-HRPEA 852 EUR
USBI041552-HRP
Anti-SENP6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100; Immunofluorescence
Type: Primary
Antigen: SENP6 (SUMO1/sentrin specific peptidase 6)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human