Stampa…
Anti-SENP6 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBI041552-BIOTINEA 852 EUR
USBI041552-BIOTIN
Anti-SENP6 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100; Immunofluorescence
Type: Primary
Antigen: SENP6 (SUMO1/sentrin specific peptidase 6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human