Stampa…

Anti-NGB Rabbit Polyclonal Antibody (Biotin)

Fornitore: US Biological
USBI038952-BIOTINEA 852 EUR
USBI038952-BIOTIN
Anti-NGB Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: NGB (neuroglobin)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR