Stampa…

Anti-NFKBIL1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: US Biological
USBI038945-PEEA 852 EUR
USBI038945-PE
Anti-NFKBIL1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: NFKBIL1 (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR