Stampa…

Anti-NFKBIA Rabbit Polyclonal Antibody (Biotin)

Fornitore: US Biological
USBI038940-BIOTINEA 852 EUR
USBI038940-BIOTIN
Anti-NFKBIA Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Dot blot 1:500

Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR