Stampa…

Anti-NFIL3 Rabbit Polyclonal Antibody

Fornitore: US Biological
USBI038935EA 587 EUR
USBI038935
Anti-NFIL3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NFIL3 (nuclear factor, interleukin 3 regulated)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR