Stampa…

Anti-NF kappa B Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: US Biological
USBI038926-APCEA 852 EUR
USBI038926-APC
Anti-NF kappa B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Dot blot 1:100-500
Do not freeze!

Type: Primary
Antigen: NF kappa B
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR