Stampa…

Anti-NEUROD6 Rabbit Polyclonal Antibody (Biotin)

Fornitore: US Biological
USBI038912-BIOTINEA 852 EUR
USBI038912-BIOTIN
Anti-NEUROD6 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NEUROD6 (neuronal differentiation 6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR