Stampa…

Anti-NEUROD1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: US Biological
USBI038911-APCEA 852 EUR
USBI038911-APC
Anti-NEUROD1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:100
Do not freeze!

Type: Primary
Antigen: NEUROD1 (neuronal differentiation 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR