Stampa…

Anti-NEU2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: US Biological
USBI038904-APCEA 852 EUR
USBI038904-APC
Anti-NEU2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NEU2 (sialidase 2 (cytosolic sialidase))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR