Stampa…

Anti-NEK4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornitore: US Biological
USBI038886-APCEA 852 EUR
USBI038886-APC
Anti-NEK4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NEK4 (NIMA-related kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR