Stampa…

Anti-NEK3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornitore: US Biological
USBI038885-APEA 852 EUR
USBI038885-AP
Anti-NEK3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NEK3 (NIMA-related kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR