Stampa…
Anti-NDUF3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornitore: US Biological
USBI038855-APEA 852 EUR
USBI038855-AP
Anti-NDUF3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: NDUF3
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse