Stampa…

Anti-NCOA7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: US Biological
USBI038837-HRPEA 852 EUR
USBI038837-HRP
Anti-NCOA7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NCOA7 (nuclear receptor coactivator 7)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR