Stampa…

Anti-NCCRP1 Rabbit Polyclonal Antibody

Fornitore: US Biological
USBI038826EA 587 EUR
USBI038826
Anti-NCCRP1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NCCRP1 (non-specific cytotoxic cell receptor protein 1 homolog (zebrafish))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR