Stampa…

Anti-NCBP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: US Biological
USBI038825-HRPEA 852 EUR
USBI038825-HRP
Anti-NCBP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NCBP1 (nuclear cap binding protein subunit 1, 80kDa)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR