Stampa…

Anti-NCAPG2 Rabbit Polyclonal Antibody

Fornitore: US Biological
USBI038823EA 587 EUR
USBI038823
Anti-NCAPG2 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NCAPG2 (non-SMC condensin II complex, subunit G2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR