Stampa…

Anti-NASP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: US Biological
USBI038801-FITCEA 852 EUR
USBI038801-FITC
Anti-NASP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NASP (nuclear autoantigenic sperm protein (histone-binding))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR