Stampa…
Anti-MTHFR Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornitore: US Biological
USBI038656-APCEA 852 EUR
USBI038656-APC
Anti-MTHFR Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTHFR (methylenetetrahydrofolate reductase (NAD(P)H))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human