Stampa…
Anti-MTHFD2L Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: US Biological
USBI038655-PEEA 852 EUR
USBI038655-PE
Anti-MTHFD2L Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTHFD2L (methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human