Stampa…
Anti-MTCH2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: US Biological
USBI038651-PEEA 852 EUR
USBI038651-PE
Anti-MTCH2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: MTCH2 (mitochondrial carrier 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse