Stampa…

Anti-MTAP Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: US Biological
USBI038650-PEEA 852 EUR
USBI038650-PE
Anti-MTAP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MTAP (methylthioadenosine phosphorylase)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR