Stampa…
Anti-MTA1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornitore: US Biological
USBI038648-FITCEA 852 EUR
USBI038648-FITC
Anti-MTA1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTA1 (metastasis associated 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human