Stampa…
Anti-MT-CO3 Rabbit Polyclonal Antibody
Fornitore: US Biological
USBI038647EA 587 EUR
USBI038647
Anti-MT-CO3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MT-CO3 (mitochondrially encoded cytochrome c oxidase III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human