Stampa…
Anti-MSX2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: US Biological
USBI038646-PEEA 852 EUR
USBI038646-PE
Anti-MSX2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MSX2 (msh homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human