Stampa…
Anti-Mst4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: US Biological
USBI038643-PEEA 852 EUR
USBI038643-PE
Anti-Mst4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: Mst4
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse