Stampa…
Anti-Mst4 Rabbit Polyclonal Antibody (Biotin)
Fornitore: US Biological
USBI038643-BIOTINEA 852 EUR
USBI038643-BIOTIN
Anti-Mst4 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: Mst4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse