Stampa…

Anti-MST1R Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: US Biological
USBI038642-PEEA 852 EUR
USBI038642-PE
Anti-MST1R Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MST1R (macrophage stimulating 1 receptor (c-met-related tyrosine kinase))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR