Stampa…
Anti-HLA-DPA1 Rabbit Polyclonal Antibody (Biotin)
Fornitore: US Biological
USBI036620-BIOTINEA 852 EUR
USBI036620-BIOTIN
Anti-HLA-DPA1 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HLA-DPA1 (major histocompatibility complex, class II, DP alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human