Italy
Order Entry
Anti-ANAPC5 Rabbit Polyclonal Antibody
  USBI031867
undefined
Anti-ANAPC5 Rabbit Polyclonal Antibody
  USBI031867
 
667,00 €
   
  1 * 200 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Anaphase Promoting Complex Subunit 5
  • Antigen symbol:
    ANAPC5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Amino acid number:
    156 to 185
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: ANAPC5 (anaphase promoting complex subunit 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human