Anti-Tau (TAU-5) Mouse Monoclonal Antibody [clone: TAU-5]
577801-100
- Antibody type:Primary
- Antigen symbol:TAU
- Clonality:Monoclonal
- Clone:TAU-5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Tau (TAU-5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: