Stampa…
Anti-SCP3 Rabbit Polyclonal Antibody
Fornitore: Thermo Fisher Scientific
PIEAPA116764EA 0 EUR
PIEAPA116764
Anti-SCP3 Rabbit Polyclonal Antibody
Anticorpi
The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis. It is the result of chromosome pairing interactions and not the cause of them.
Type: Primary
Antigen: SCP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Mouse, Porcine