Suggested positive control: antigen standard for ADNP (transient overexpression lysate). The activity-dependent neuroprotective protein (hADNP) gene is frequently amplified in many neoplasias, including breast, bladder, ovarian, pancreatic, and colon cancers. hADNP mRNA is abundantly expressed in distinct normal tissues, and high expression levels were encountered in malignant cells. hADNP is implicated in maintaining cell survival, perhaps through modulation of p53.
Type: Primary
Antigen: ADNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human