Stampa…
Anti-Kallikrein 4 Rabbit Polyclonal Antibody
Fornitore: Thermo Fisher Scientific
PIEAPA116651EA 0 EUR
PIEAPA116651
Anti-Kallikrein 4 Rabbit Polyclonal Antibody
Anticorpi
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
Type: Primary
Antigen: Kallikrein 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human