Keep my session open?
Ending In 
The session is expired
La tua sessione è scaduta. Per favore rifare il login

Anti-LYVE1 Rabbit Polyclonal Antibody

Fornitore: Thermo Scientific
Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: mouse spleen lysate or protein. The lymphatic vasculature forms a second circulatory system that drains extracellular fluid from the tissues and provides an exclusive environment in which immune cells can encounter and respond to foreign antigen. Recently a number of interesting molecules have been identified that may be exploited as markers for lymphatic endothelium, including the hyaluronan receptor LYVE1, PALE, VEGFR3, and podoplanin. LYVE1 has been identified as a major receptor for HA (extracellular matrix glycosaminoglycan hyaluronan) on the lymph vessel wall. The deduced amino acid sequence of LYVE1 predicts a 322-residue type I integral membrane polypeptide 41% similar to the CD44 HA receptor with a 212-residue extracellular domain containing a single Link module, the prototypic HA binding domain of the Link protein superfamily. Like CD44, the LYVE1 molecule binds both soluble and immobilized HA. However, unlike CD44, the LYVE1 molecule colocalizes with HA on the luminal face of the lymph vessel wall and is completely absent from blood vessels. Hence, LYVE1 is the first lymph-specific HA receptor to be characterized and is a uniquely powerful marker for lymph vessels themselves.

Type: Primary
Antigen: LYVE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse




 

Ordine

Espandi tutto / Chiudi tutto
Taglia Conf Codice catalogo VWR Quantità
100 µL 100 µl PIEAPA116635 Blocked
Codice articolo fornitore: PA116635
Codice articolo locale: PIEAPA116635
Blocked Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.

Specifiche

Antigene LYVE1
Nome antigene Extracellular link Domain Containing 1
Reattività Human, Mouse, Rat
Coniugazione Unconjugated
Clonabilità Polyclonal
Tipo anticorpo Primary
Host Rabbit
Isotipo IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Sinomini antigene HAR, LYVE-1, lymphatic vessel endothelial hyaluronan receptor 1, XLKD1, Extracellular link domain containing 1, hyaluronic acid receptor
ID gene 293186, 10894, 114332
Concentrazione 1 mg/ml
Immunogeno A synthetic peptide made to a C-terminal portion of the murine LYVE-1 protein sequence (between residues 250-318).
Purificazione Antigen affinity chromatography
Forma Liquid
Tampone di conservazione PBS, 0.02% sodium azide
Temperatura di conservazione Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Temperatura di spedizione Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Seleziona tra i certificati dei lotti più recenti:

o
Confronta le alternative
Abbiamo individuato delle alternative che potrebbero farti risparmiare  per articolo.  Per confrontare gli articoli, puoi selezionare fino a 3 alternative proposte e cliccare "Confronta". Per aggiungerli al carrello inserisci una quantità e clicca "Aggiungi al carrello".

Prodotto originariamente selezionato:

Descrizione Codice catalogo Disponibilità Unità Il tuo prezzo Prezzo per Quantità