Stampa…

Anti-CTGF Rabbit Polyclonal Antibody

Fornitore: Thermo Fisher Scientific
PIEAPA116634EA 0 EUR
PIEAPA116634
Anti-CTGF Rabbit Polyclonal Antibody
Anticorpi
This antibody is predicted to react with porcine, bovine, chicken and sheep based on 100% sequence homology. Suggested positive control: mouse kidney lysate or protein. CTGF (connective tissue growth factor) is a 38kDa, cysteine-rich, secreted peptide. It is a new member of the peptide family that include serum-induced immediate early gene products, a v-src-induced peptide and a putative proto-oncogene. Among the many functions of the CTGF gene family are embryogenesis, wound healing and regulation of extracellular matrix production.

Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse, Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR