Keep my session open?
Ending In 
The session is expired
La tua sessione è scaduta. Per favore rifare il login

Anti-HIF1A Rabbit Polyclonal Antibody

Fornitore: Thermo Scientific
This antibody has 93% sequence homology with bovine and 90% sequence homology with mouse. Suggested positive control: Cos-7 Hypoxia induced nuclear extract, COS-7 nuclear extracts (treated). Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease. HIF-1 is a nuclear protein involved in mammalian oxygen homeostasis. This occurs as a posttranslational modification by prolyl hydroxylation. HIF-1 is a heterodimer composed of HIF-1 alpha and HIF-1 beta subunits. Both subunits are constantly translated. However, under normoxic conditions, human HIF-1 alpha is hydroxylated at Pro402 or Pro564 by a set of HIF prolyl hydroxylases, is polyubiquinated, and eventually degraded in proteosomes. Under hypoxic conditions, the lack of hydroxylation prevents HIF degradation and increases transcriptional activity. Therefore, the concentration of HIF-1 alpha increases in the cell. In contrast, HIF-1 beta remains stable under either condition. HIF hydroxylases provide insight into hypoxic cell responses, which may be used to help isolate therapeutic targets.

Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine




 

Ordine

Espandi tutto / Chiudi tutto
Taglia Conf Codice catalogo VWR Quantità
100 µL 100 µl PIEAPA116627 Blocked
Codice articolo fornitore: PA116627
Codice articolo locale: PIEAPA116627
Blocked Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.

Specifiche

Antigene HIF1A
Nome antigene HIF1 alpha
Reattività Rat, Porcine, Human, Mouse
Coniugazione Unconjugated
Clonabilità Polyclonal
Tipo anticorpo Primary
Host Rabbit
Isotipo IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Sinomini antigene Hypoxia Inducible Factor 1 alpha, MOP1, PASD8, ARNT2, ARNT interacting protein
ID gene 15251, 3091, 29560, 396696
Concentrazione 1 mg/ml
Immunogeno Fusion protein containing amino acids 432-528 of human HIF-1alpha
Purificazione Antigen affinity chromatography
Forma Liquid
Tampone di conservazione tris glycine, 0.15M NaCl, 0.05% sodium azide
Temperatura di conservazione -20° C, Avoid Freeze/Thaw Cycles
Temperatura di spedizione Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Seleziona tra i certificati dei lotti più recenti:

o
Confronta le alternative
Abbiamo individuato delle alternative che potrebbero farti risparmiare  per articolo.  Per confrontare gli articoli, puoi selezionare fino a 3 alternative proposte e cliccare "Confronta". Per aggiungerli al carrello inserisci una quantità e clicca "Aggiungi al carrello".

Prodotto originariamente selezionato:

Descrizione Codice catalogo Disponibilità Unità Il tuo prezzo Prezzo per Quantità