Stampa…
Anti-Tankyrase Rabbit Polyclonal Antibody
Fornitore: Thermo Fisher Scientific
PIEAPA116550EA 0 EUR
PIEAPA116550
Anti-Tankyrase Rabbit Polyclonal Antibody
Anticorpi
In Western blot, this antibody recognizes an endogenous band at ~150 kDa in whole cell extract and a band at ~180 kDa in baculovirus-derived his-tagged protein. Suggested positive control: Hela whole cell extract. The ~142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintenance of telomere length.
Type: Primary
Antigen: Tankyrase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human