Stampa…
Anti-TRAPPC2 Rabbit Monoclonal Antibody [clone: EPR3401]
Fornitore: Abcam
ABCAAB111848-100EA 875 EUR
ABCAAB111848-100
Anti-TRAPPC2 Rabbit Monoclonal Antibody [clone: EPR3401]
Anticorpi
Type: Primary
Antigen: TRAPPC2 (trafficking protein particle complex 2)
Clonality: monoclonal
Clone: EPR3401
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici