Stampa…
Anti-PHF6 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB111844-50EA 697 EUR
ABCAAB111844-50
Anti-PHF6 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PHF6 (PHD finger protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human