Stampa…
Anti-RAF1 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB111829-100EA 882 EUR
ABCAAB111829-100
Anti-RAF1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human