Stampa…
Anti-SLC7A11 Goat Polyclonal Antibody
Fornitore: Abcam
ABCAAB111822-50EA 713 EUR
ABCAAB111822-50
Anti-SLC7A11 Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human