Stampa…

Anti-SAR1 Goat polyclonal antibody unconjugated

ABCAAB111814-50EA 682 EUR
ABCAAB111814-50
Anti-SAR1 Goat polyclonal antibody unconjugated
Type: Primary
Antigen: SAR1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR