Stampa…

Anti-EXOSC2 Rabbit polyclonal antibody unconjugated

Fornitore: Abcam
ABCAAB111810-50EA 697 EUR
ABCAAB111810-50
Anti-EXOSC2 Rabbit polyclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: EXOSC2 (exosome component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR