Stampa…
Anti-CYP4F2 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB111741-100EA 713 EUR
ABCAAB111741-100
Anti-CYP4F2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CYP4F2 (cytochrome P450, family 4, subfamily F, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human