Stampa…

Anti-SAP130 Rabbit Polyclonal Antibody

Fornitore: Abcam
ABCAAB111739-50EA 902 EUR
ABCAAB111739-50
Anti-SAP130 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: SAP130 (Sin3A-associated protein, 130kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR