Stampa…
Anti-FUBP1 Rabbit polyclonal antibody unconjugated
ABCAAB111738-50EA 672 EUR
ABCAAB111738-50
Anti-FUBP1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: FUBP1 (far upstream element (FUSE) binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human