Stampa…

Anti-MRPS10 Rabbit Polyclonal Antibody

Fornitore: Abcam
ABCAAB111715-50EA 665 EUR
ABCAAB111715-50
Anti-MRPS10 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MRPS10 (mitochondrial ribosomal protein S10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR