Stampa…
Anti-MARC2 Rabbit Polyclonal Antibody
Fornitore: Abcam
ABCAAB111685-50EA 837 EUR
ABCAAB111685-50
Anti-MARC2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MARC2 (mitochondrial amidoxime reducing component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human